Learn More
Cayman Chemical NDSethylamIodaronhydroch 100mg
SDP
Catalog No. 502893622
Encompass_Preferred
the major CYP3A4 metabolite of amiodarone; used as an analytical reference standard for monitoring clinical efficacy of amiodarone in plasma samples
Catalog No. 50-289-3622 Supplier Cayman Chemical Supplier No. 9000537100 MG
the major CYP3A4 metabolite of amiodarone; used as an analytical reference standard for monitoring clinical efficacy of amiodarone in plasma samples