Learn More
Cayman Chemical N-desmethyl Levofloxacin (hydrochloride), Purity: ≥98%, 10 mg
SDP
Catalog No. 502939565
Encompass_Preferred
An active metabolite of levofloxacin; active against S. aureus, S. epidermidis, B. subtilis, E. coli, P. aeruginosa, and K. pneumoniae (MICs = 4, 1, 1, 0.012, >4, and 0.25 μg/ml, respectively)
Catalog No. 50-293-9565 Supplier Cayman Chemical Supplier No. 2717810 MG
An active metabolite of levofloxacin; active against S. aureus, S. epidermidis, B. subtilis, E. coli, P. aeruginosa, and K. pneumoniae (MICs = 4, 1, 1, 0.012, >4, and 0.25 μg/ml, respectively)