Learn More
Aviva Systems Biology Corp NDUFA1 Antibody - N-terminal region (ARP64705_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64705_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NDUFA1. It was validated on Western Blot. . Alternate names: CI-MWFE, MWFE, ZNF183ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-288-341