Learn More
Aviva Systems Biology Corp NDUFA6 Antibody - C-terminal region (ARP62484_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62484_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NDUFA6. It was validated on Western Blot. . Alternate names: B14, CI-B14, LYRM6, NADHB14ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-694