Learn More
Aviva Systems Biology Corp NDUFA8 Antibody - N-terminal region (ARP62418_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62418_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NDUFA8. It was validated on Western Blot. . Alternate names: CI-19KD, CI-PGIV, MGC793, PGIVApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-676