Learn More
ABclonal Technology NDUFAB1 Rabbit pAb
SDP
Catalog No. 5000104653
Encompass_Preferred
Predicted to enable acyl binding activity, acyl carrier activity, and fatty acid binding activity. Involved in mitochondrial respiratory chain complex I assembly and protein lipoylation. Located in mitochondrion and nucleoplasm. Part of mitochondrial respiratory chain complex I. Colocalizes with mitochondrial large ribosomal subunit.
Catalog No. 50-001-04653 Supplier ABclonal Technology Supplier No. A14657500UL
Predicted to enable acyl binding activity, acyl carrier activity, and fatty acid binding activity. Involved in mitochondrial respiratory chain complex I assembly and protein lipoylation. Located in mitochondrion and nucleoplasm. Part of mitochondrial respiratory chain complex I. Colocalizes with mitochondrial large ribosomal subunit.