Learn More
Aviva Systems Biology Corp NDUFB10 Antibody - C-terminal region (ARP62883_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62883_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NDUFB10. It was validated on Western Blot. . Alternate names: PDSWApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-287-799