Learn More
ABclonal Technology NDUFB11 Rabbit pAb
SDP
Catalog No. 5000108102
Encompass_Preferred
The protein encoded by this gene is a subunit of the multisubunit NADHubiquinone oxidoreductase (complex I). Mammalian complex I is located at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to ubiquinone. Mutations in the human gene are associated with linear skin defects with multiple congenital anomalies 3 and mitochondrial complex I deficiency.
Catalog No. 50-001-08102 Supplier ABclonal Technology Supplier No. A15617500UL
The protein encoded by this gene is a subunit of the multisubunit NADHubiquinone oxidoreductase (complex I). Mammalian complex I is located at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to ubiquinone. Mutations in the human gene are associated with linear skin defects with multiple congenital anomalies 3 and mitochondrial complex I deficiency.