Learn More
ABclonal Technology NDUFB3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A143781000UL
Encompass_Preferred
This gene encodes an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) which is the first enzyme in the electron transport chain of mitochondria. This protein localizes to the inner membrane of the mitochondrion as a single-pass membrane protein. Mutations in this gene contribute to mitochondrial complex 1 deficiency. Alternative splicing results in multiple transcript variants encoding the same protein. Humans have multiple pseudogenes of this gene.
Catalog No. 50-001-08267