Learn More
Aviva Systems Biology Corp NDUFB7 Antibody - N-terminal region (ARP62290_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62290_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NDUFB7. It was validated on Western Blot. . Alternate names: B18, CI-B18, MGC2480ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-287-648