Learn More
Aviva Systems Biology Corp Ndufb8 Antibody - C-terminal region (ARP46559_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46559_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ndufb8. It was validated on Western Blot. . Alternate names: 2900010I05Rik, AI987932ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-264