Learn More
Aviva Systems Biology Corp NDUFC1 Antibody - middle region (ARP56360_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56360_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NDUFC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NDUFC1. Alternate names: KFYI, MGC117464, MGC126847, MGC138266ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61829