Learn More
Aviva Systems Biology Corp NDUFS1 Antibody - middle region (ARP56609_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56609_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NDUFS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NDUFS1. Alternate names: CI-75Kd, MGC26839, PRO1304, CI-75kApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62860