Learn More
Aviva Systems Biology Corp Ndufs6 Antibody - N-terminal region (ARP56580_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56580_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ndufs6. It was validated on Western Blot. . Alternate names: BC059730, IP13, MGC107676ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-179