Learn More
Aviva Systems Biology Corp NDUFS8 Antibody - C-terminal region (ARP61825_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61825_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NDUFS8. It was validated on Western Blot. . Alternate names: CI23KD, TYKY, CI-23kApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-287-479