Learn More
Aviva Systems Biology Corp Ndufv2 Antibody - C-terminal region (ARP57510_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57510_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ndufv2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human Ndufv2. Alternate names: 2900010C23RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-327