Learn More
Medchemexpress LLC HY-16011 5mg Medchemexpress, NE 10790 CAS:152831-36-2 Purity:>98%
SDP
Catalog No. 501932760
Encompass_Preferred
Medchemexpress, HY-16011 5mg NE 10790 CAS:152831-36-2 NE 10790, a poor farnesyl pyrophosphate synthase inhibitor, is a phosphonocarboxylate analogue of the potent bisphosphonate risedronate and is a weak antiresorptive agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2760 Supplier Medchemexpress LLC Supplier No. HY160115MG
Medchemexpress, HY-16011 5mg NE 10790 CAS:152831-36-2 NE 10790, a poor farnesyl pyrophosphate synthase inhibitor, is a phosphonocarboxylate analogue of the potent bisphosphonate risedronate and is a weak antiresorptive agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.