Learn More
Electron Microscopy Sciences Concentric Circles Microscope Graticle NE43, 21 mm, 10 circles 0.5 mm-5 mm
SDP
Catalog No. 5025095
Encompass_Preferred
Can be used for two-way measurement when calibrated as a micrometer. There are 10 concentric circles 0.5mm-5mm in diameter. Surface chrome image.
Size: 21 mm
Catalog No. 50-250-95 Supplier Electron Microscopy Sciences Supplier No. 6799721
Can be used for two-way measurement when calibrated as a micrometer. There are 10 concentric circles 0.5mm-5mm in diameter. Surface chrome image.
Size: 21 mm