Learn More
Medchemexpress LLC HY-101784 100mg Medchemexpress, NE 52-QQ57 CAS:1401728-56-0 Purity:>98%
SDP
Catalog No. 501932214
Encompass_Preferred
Medchemexpress, HY-101784 100mg NE 52-QQ57 CAS:1401728-56-0 NE 52-QQ57 is a selective, and orally available GPR4 antagonist with an IC 50 of 70 nM [1] . Anti-inflammatory activities [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2214 Supplier Medchemexpress LLC Supplier No. HY101784100MG
Medchemexpress, HY-101784 100mg NE 52-QQ57 CAS:1401728-56-0 NE 52-QQ57 is a selective, and orally available GPR4 antagonist with an IC 50 of 70 nM [1] . Anti-inflammatory activities [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.