Learn More
Cayman Chemical Nε,Nε,Nε-Trimethyllysine (chloride), Purity: ≥95%, CAS: 55528-53-5, 25 mg
SDP
Catalog No. 502947378
Encompass_Preferred
An amino acid and a precursor in L-carnitine biosynthesis; produced by intestinal microbiota or can be obtained by the degradation of post-translationally modified proteins or dietary sources
Catalog No. 50-294-7378 Supplier Cayman Chemical Supplier No. 3389825 MG
An amino acid and a precursor in L-carnitine biosynthesis; produced by intestinal microbiota or can be obtained by the degradation of post-translationally modified proteins or dietary sources