Learn More
Electron Microscopy Sciences Square Grid Graticle NE10, 16mm, 0.5 mm pitch
SDP
Catalog No. 5028861
Encompass_Preferred
Simple grids are convenient for making sketches of the obseved specimen on graph paper. They are also useful for particle counting. Grid (net) 0.5mm pitch.
Size: 16mm
Divisions: 0.5 mm pitch
Surface: Chrome image
Catalog No. 50-288-61 Supplier Electron Microscopy Sciences Supplier No. 6801116
Simple grids are convenient for making sketches of the obseved specimen on graph paper. They are also useful for particle counting. Grid (net) 0.5mm pitch.
Size: 16mm
Divisions: 0.5 mm pitch
Surface: Chrome image