Learn More
Electron Microscopy Sciences Square Grid Graticle NE11, 16mm, 1.00 mm pitch
SDP
Catalog No. 5028863
Encompass_Preferred
Simple grids are convenient for making sketches of the obseved specimen on graph paper. They are also useful for particle counting. Grid (net) 1.00mm pitch.
Size: 16mm
Divisions: 1.00 mm pitch
Surface: Chrome image
Catalog No. 50-288-63 Supplier Electron Microscopy Sciences Supplier No. 6801516
Simple grids are convenient for making sketches of the obseved specimen on graph paper. They are also useful for particle counting. Grid (net) 1.00mm pitch.
Size: 16mm
Divisions: 1.00 mm pitch
Surface: Chrome image