Learn More
Electron Microscopy Sciences Square Grid Graticle NE11, 21mm, 1.00 mm pitch
SDP
Catalog No. 5028865
Encompass_Preferred
Simple grids are convenient for making sketches of the obseved specimen on graph paper. They are also useful for particle counting.
Size: 21mm
Divisions: 1.00 mm pitch
Surface: Chrome image
Catalog No. 50-288-65 Supplier Electron Microscopy Sciences Supplier No. 6801521
Simple grids are convenient for making sketches of the obseved specimen on graph paper. They are also useful for particle counting.
Size: 21mm
Divisions: 1.00 mm pitch
Surface: Chrome image