Learn More
Electron Microscopy Sciences Chessboard Squares Graticle NE15, 16mm
SDP
Catalog No. 5031204
Encompass_Preferred
The dark squares are translucent. Used as an alternative to simple grids for area of specimen determination and paricle counting. Alternative light and dark squares help to reduce eyestrain.
Size: 16mm
Divisions: 2.0 mm squares
Catalog No. 50-312-04 Supplier Electron Microscopy Sciences Supplier No. 6800816
The dark squares are translucent. Used as an alternative to simple grids for area of specimen determination and paricle counting. Alternative light and dark squares help to reduce eyestrain.
Size: 16mm
Divisions: 2.0 mm squares