Learn More
Electron Microscopy Sciences Chessboard Squares Graticle NE15, 19mm
SDP
Catalog No. 5031203
Encompass_Preferred
The dark squares are translucent. Used as an alternative to simple grids for area of specimen determination and paricle counting. Alternative light and dark squares help to reduce eyestrain.
Size: 19mm
Divisions: 2.0 mm squares
Catalog No. 50-312-03 Supplier Electron Microscopy Sciences Supplier No. 6800819
The dark squares are translucent. Used as an alternative to simple grids for area of specimen determination and paricle counting. Alternative light and dark squares help to reduce eyestrain.
Size: 19mm
Divisions: 2.0 mm squares