Learn More
Electron Microscopy Sciences Gauge Pairs NE19, 16mm
SDP
Catalog No. 5031213
Encompass_Preferred
Gauge pairs occupying a field of view of 10 mm. Each gauge is proportional to its adjacent number. The approximately size of the smallest pair = 0.1 mm.
Size: 16 mm
Catalog No. 50-312-13 Supplier Electron Microscopy Sciences Supplier No. 6800116
Gauge pairs occupying a field of view of 10 mm. Each gauge is proportional to its adjacent number. The approximately size of the smallest pair = 0.1 mm.
Size: 16 mm