Learn More
Electron Microscopy Sciences Gauge Pairs NE19, 19mm
SDP
Catalog No. 5031212
Encompass_Preferred
Gauge pairs occupying a field of view of 10 mm. Each gauge is proportional to its adjacent number. The approximately size of the smallest pair = 0.1 mm.
Size: 19 mm
Catalog No. 50-312-12 Supplier Electron Microscopy Sciences Supplier No. 6800119
Gauge pairs occupying a field of view of 10 mm. Each gauge is proportional to its adjacent number. The approximately size of the smallest pair = 0.1 mm.
Size: 19 mm