Learn More
Electron Microscopy Sciences Concentric Circles Graticle NE22, 16mm, 0.5 mm - 12 mm diameter
SDP
Catalog No. 5031210
Encompass_Preferred
This design leaves the circle clear of obstruction. In addition the intermediate lines are broken to improve ease of reading.
Size: 16mm
Catalog No. 50-312-10 Supplier Electron Microscopy Sciences Supplier No. 6800016
This design leaves the circle clear of obstruction. In addition the intermediate lines are broken to improve ease of reading.
Size: 16mm