Learn More
Electron Microscopy Sciences Square Grid Graticle NE34, 16mm, 0.1 mm squares
SDP
Catalog No. 5028869
Encompass_Preferred
Simple grids are convenient for making sketches of the obseved specimen on graph paper. They are also useful for particle counting.
Size: 16mm
Divisions: 0.1 mm squares
Surface: Chrome image
Catalog No. 50-288-69 Supplier Electron Microscopy Sciences Supplier No. 6801816
Simple grids are convenient for making sketches of the obseved specimen on graph paper. They are also useful for particle counting.
Size: 16mm
Divisions: 0.1 mm squares
Surface: Chrome image