Learn More
Electron Microscopy Sciences Square Grid Graticle NE34, 21mm, 0.1 mm squares
SDP
Catalog No. 5028871
Encompass_Preferred
Simple grids are convenient for making sketches of the obseved specimen on graph paper. They are also useful for particle counting.
Size: 21mm
Divisions: 0.1 mm squares
Surface: Chrome image
Catalog No. 50-288-71 Supplier Electron Microscopy Sciences Supplier No. 6801821
Simple grids are convenient for making sketches of the obseved specimen on graph paper. They are also useful for particle counting.
Size: 21mm
Divisions: 0.1 mm squares
Surface: Chrome image