Learn More
Electron Microscopy Sciences Indexed Grid Microscope Graticle NE35, 21mm, 1 mm indexed squares
SDP
Catalog No. 5026652
Encompass_Preferred
Useful for area of specimen determinations, especially rectangular shapes, also for particle counting.
Size: 21mm
Divisions: 1 mm indexed squares
Surface: Chrome image
Catalog No. 50-266-52 Supplier Electron Microscopy Sciences Supplier No. 6801421
Useful for area of specimen determinations, especially rectangular shapes, also for particle counting.
Size: 21mm
Divisions: 1 mm indexed squares
Surface: Chrome image