Learn More
Electron Microscopy Sciences Squares Graticle NE38, 19mm, 10 mm, 7 mm and 4 mm squares
SDP
Catalog No. 5028965
Encompass_Preferred
Combines three areas in one for convenience, giving area ratios A:B of 1:3 and B:C of 1:2.
Size: 19mm
Divisions: 10 mm, 7 mm and 4 mm squares
Surface: Chrome image
Catalog No. 50-289-65 Supplier Electron Microscopy Sciences Supplier No. 6801719
Combines three areas in one for convenience, giving area ratios A:B of 1:3 and B:C of 1:2.
Size: 19mm
Divisions: 10 mm, 7 mm and 4 mm squares
Surface: Chrome image