Learn More
Electron Microscopy Sciences Concentric Circles Microscope Graticle NE44, 21 mm, 10 circles 1 mm-10 mm
SDP
Catalog No. 5025098
Encompass_Preferred
Can be used for two-way measurement when calibrated as a micrometer. There are 10 concentric circles 1mm-10mm in diameter. Surface chrome image.
Size: 21 mm
Catalog No. 50-250-98 Supplier Electron Microscopy Sciences Supplier No. 6799821
Can be used for two-way measurement when calibrated as a micrometer. There are 10 concentric circles 1mm-10mm in diameter. Surface chrome image.
Size: 21 mm