Learn More
Electron Microscopy Sciences Concentric Circles Microscope Graticle NE44, 26 mm, 10 circles 1 mm-10 mm
SDP
Catalog No. 501928020
Encompass_Preferred
Can be used for two-way measurement when calibrated as a micrometer. There are 10 concentric circles 1mm-10mm in diameter. Surface chrome image.
Size: 26 mm
Catalog No. 50-192-8020 Supplier Electron Microscopy Sciences Supplier No. 6799826
Add to Cart
Add to Cart
Can be used for two-way measurement when calibrated as a micrometer. There are 10 concentric circles 1mm-10mm in diameter. Surface chrome image.
Size: 26 mm