Learn More
Electron Microscopy Sciences Eyepiece Scale NE45, 16mm
SDP
Catalog No. 5031214
Encompass_Preferred
Full protractor scale is 10 mm in diameter and divided in degrees. Surface chrome image.
Size: 16 mm
Catalog No. 50-312-14 Supplier Electron Microscopy Sciences Supplier No. 6800216
Full protractor scale is 10 mm in diameter and divided in degrees. Surface chrome image.
Size: 16 mm