Learn More
Electron Microscopy Sciences Eyepiece Scale NE45, 19mm
SDP
Catalog No. 5031216
Encompass_Preferred
Full protractor scale is 10 mm in diameter and divided in degrees. Surface chrome image.
Size: 19 mm
Catalog No. 50-312-16 Supplier Electron Microscopy Sciences Supplier No. 6800219
Full protractor scale is 10 mm in diameter and divided in degrees. Surface chrome image.
Size: 19 mm