Learn More
Electron Microscopy Sciences Concentric Circles Microscope Graticle NE47, 19 mm, 10 circles 2 mm-20 mm
SDP
Catalog No. 5025099
Encompass_Preferred
Can be used for two-way measurement when calibrated as a micrometer. There are 10 concentric circles 1mm-10mm in diameter. Surface chrome image.
Size: 19 mm
Catalog No. 50-250-99 Supplier Electron Microscopy Sciences Supplier No. 6799919
Can be used for two-way measurement when calibrated as a micrometer. There are 10 concentric circles 1mm-10mm in diameter. Surface chrome image.
Size: 19 mm