Learn More
Electron Microscopy Sciences Concentric Circles Microscope Graticle NE47, 21 mm, 10 circles 2 mm-20 mm
SDP
Catalog No. 5025092
Encompass_Preferred
Can be used for two-way measurement when calibrated as a micrometer. There are 10 concentric circles 2mm-20mm in diameter. Surface chrome image.
Size: 21 mm
Catalog No. 50-250-92 Supplier Electron Microscopy Sciences Supplier No. 6799921
Can be used for two-way measurement when calibrated as a micrometer. There are 10 concentric circles 2mm-20mm in diameter. Surface chrome image.
Size: 21 mm