Learn More
Electron Microscopy Sciences Single Line Graticle NE50, 19mm, 0.02 mm Line width
SDP
Catalog No. 5028633
Encompass_Preferred
For measurement of large objects and alignment in conjunction with a graduated mechanical stage. Single line, nominal width 0.02 mm, image covers entire field of view, surface chrome image.
Size: 19mm
Line width: 0.02 mm
Surface: Chrome image
Catalog No. 50-286-33 Supplier Electron Microscopy Sciences Supplier No. 6798219
For measurement of large objects and alignment in conjunction with a graduated mechanical stage. Single line, nominal width 0.02 mm, image covers entire field of view, surface chrome image.
Size: 19mm
Line width: 0.02 mm
Surface: Chrome image