Learn More
Electron Microscopy Sciences Broken Crosslines Microscope Graticle NE56, 21mm, Broken line
SDP
Catalog No. 5024790
Encompass_Preferred
For measurement of large objects in conjunction with a graduated mechanical stage. Measurements in two directions and for sighting and alignment. Broken lines enable fine detail to be seen at the breaks. A thin boundary would be lost behind a a continuous line. Image covers the entire field of view. Surface chrome image.
Size: 21mm
Divisions: Broken line
Catalog No. 50-247-90 Supplier Electron Microscopy Sciences Supplier No. 6798521
For measurement of large objects in conjunction with a graduated mechanical stage. Measurements in two directions and for sighting and alignment. Broken lines enable fine detail to be seen at the breaks. A thin boundary would be lost behind a a continuous line. Image covers the entire field of view. Surface chrome image.
Size: 21mm
Divisions: Broken line