Learn More
Electron Microscopy Sciences Scale with Cross Line Microscope Graticle NE7, 19 mm, 10 mm with 0.1 mm spacing
SDP
Catalog No. 5028492
Encompass_Preferred
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis. Horizontal micrometer scale 10mm long with crosslines at 0.1mm spacing. Surface chrome image.
Size: 19 mm
Divisions: 10 mm with 0.1 mm spacing
Surface: Chrome image
Catalog No. 50-284-92 Supplier Electron Microscopy Sciences Supplier No. 6801319
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis. Horizontal micrometer scale 10mm long with crosslines at 0.1mm spacing. Surface chrome image.
Size: 19 mm
Divisions: 10 mm with 0.1 mm spacing
Surface: Chrome image