Learn More
Electron Microscopy Sciences Scale with Cross Line Microscope Graticle NE77, 21 mm, 5 mm with 100 divisions
SDP
Catalog No. 5028494
Encompass_Preferred
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis. Horizontal micrometer scale 5mm long in 100 divisions with crosslines. Surface chrome image.
Size: 21 mm
Divisions: 5 mm with 100 divisions
Surface: Chrome image
Catalog No. 50-284-94 Supplier Electron Microscopy Sciences Supplier No. 6801221
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis. Horizontal micrometer scale 5mm long in 100 divisions with crosslines. Surface chrome image.
Size: 21 mm
Divisions: 5 mm with 100 divisions
Surface: Chrome image