Learn More
Electron Microscopy Sciences Cross Lines Graticle NE8, 19 mm, Nominal line width 0.020 mm
SDP
Catalog No. 5031197
Encompass_Preferred
Used as NE50 but for measurements in two directions and for sighting and alignment. Image covers the entire field of view.
Size: 19 mm
Divisions: Nominal line width 0.020 mm
Surface: Chrome image
Catalog No. 50-311-97 Supplier Electron Microscopy Sciences Supplier No. 6798305
Used as NE50 but for measurements in two directions and for sighting and alignment. Image covers the entire field of view.
Size: 19 mm
Divisions: Nominal line width 0.020 mm
Surface: Chrome image