Learn More
Perkin Elmer US LLC Near-Infrared Performance Validation Upgrade Kit
Supplier:  Perkin Elmer US LLC L1250475
Encompass_Preferred
Instrument Performance Validation (IPV) kits are available for both mid-IR and near-FT-IR instruments. All kits include a polystyrene Traceable Reference Material (TRM) for abscissa calibration valid for three years. This kit is the replacement for expired TRM kits.
Catalog No. 50-211-2716