Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NEAS/alpha-Fodrin Antibody (YA1355) | 10 UL
SDP

Catalog No. 5000441612
Encompass_Preferred

NEAS/alpha-Fodrin Antibody (YA1355) is a Rabbit-derived, non-conjugated IgG monoclonal antibody designed for research applications. It targets NEAS/alpha-Fodrin, a protein involved in secretion and cytoskeletal movement. This affinity-purified antibody is suitable for Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF) applications.

  • Rabbit host
  • Recombinant, monoclonal clonality
  • Endogenous sensitivity
  • Non-conjugated
  • Liquid appearance
  • Human, mouse, rat species reactivity
  • Immunogen is a synthetic peptide of human NEAS
  • Target protein involved in signal transduction
  • Subcellular localization in cytoplasm and cytoskeleton

Catalog No. 50-004-41612 Supplier Medchemexpress LLC Supplier No. HYP8161010UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NEAS/alpha-Fodrin Antibody (YA1355) is a Rabbit-derived, non-conjugated IgG monoclonal antibody designed for research applications. It targets NEAS/alpha-Fodrin, a protein involved in secretion and cytoskeletal movement. This affinity-purified antibody is suitable for Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF) applications.

  • Rabbit host
  • Recombinant, monoclonal clonality
  • Endogenous sensitivity
  • Non-conjugated
  • Liquid appearance
  • Human, mouse, rat species reactivity
  • Immunogen is a synthetic peptide of human NEAS
  • Target protein involved in signal transduction
  • Subcellular localization in cytoplasm and cytoskeleton

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.