Learn More
Medchemexpress LLC NEAS/alpha-Fodrin antibody (YA1355) | 100 UL
SDP
Catalog No. 5000440848
Encompass_Preferred
NEAS/alpha-Fodrin Antibody (YA1355) is also known by synonyms including SPTAN1, NEAS, SPTA2, Spectrin alpha chain, non-erythrocytic 1, Alpha-ll spectrin, Fodrin alpha chain, Spectrin, and non-erythroid alpha subunit. It is a rabbit-derived, non-conjugated antibody reactive with human, mouse, and rat samples. It is suitable for Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF) applications.
  • Supplied as a liquid with a concentration of 0.3 mg/mL
  • Formulated in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Predicted and observed band size of 285 kDa
  • Recommended dilution ratios for WB: 1:500-1:1000
  • Recommended dilution ratios for ICC/IF: 1:50-1:200
Catalog No. 50-004-40848 Supplier Medchemexpress LLC Supplier No. HYP81610100UL
NEAS/alpha-Fodrin Antibody (YA1355) is also known by synonyms including SPTAN1, NEAS, SPTA2, Spectrin alpha chain, non-erythrocytic 1, Alpha-ll spectrin, Fodrin alpha chain, Spectrin, and non-erythroid alpha subunit. It is a rabbit-derived, non-conjugated antibody reactive with human, mouse, and rat samples. It is suitable for Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF) applications.
  • Supplied as a liquid with a concentration of 0.3 mg/mL
  • Formulated in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Predicted and observed band size of 285 kDa
  • Recommended dilution ratios for WB: 1:500-1:1000
  • Recommended dilution ratios for ICC/IF: 1:50-1:200