Learn More
Polysciences, Inc. NEAT STAIN HEM. STAIN KIT
SDP
Supplier:  Polysciences, Inc. 250341
Encompass
Neat Stain offers the flexibility of a three dip staining sequence in both the stain and counter-stain solution. Performs 150 tests.
Catalog No. NC0476613