Learn More
Electron Microscopy Sciences Neat Stain™ Hematology Stain Kit, (3) 4oz Bottles
SDP
Catalog No. 50366852
Encompass_Preferred
This kit is designed for the differentiation of morphological cell types in peripheral blood smears. The staining characteristics are similar to the traditional Wright’s and Wright-Giemsa stains.
The simple three-step procedure is useful for laboratories performing manual staining of microorganisms and spermatozoa. The kit is also well suited for stat procedures.
Each kit contains the following components:
4 oz. bottle of QUICK III Fixative Solution
4 oz. bottle of QUICK III Solution I
4 oz. bottle of QUICK III Solution II
Catalog No. 50-366-852 Supplier Electron Microscopy Sciences Supplier No. 2710401
This kit is designed for the differentiation of morphological cell types in peripheral blood smears. The staining characteristics are similar to the traditional Wright’s and Wright-Giemsa stains.
The simple three-step procedure is useful for laboratories performing manual staining of microorganisms and spermatozoa. The kit is also well suited for stat procedures.
Each kit contains the following components:
4 oz. bottle of QUICK III Fixative Solution
4 oz. bottle of QUICK III Solution I
4 oz. bottle of QUICK III Solution II