Learn More
New England Biolabs, Inc. NEB® 5-alpha Competent E. coli (High Efficiency) – 1 x 384 well plate (10 µl/well)
SDP
Catalog No. 502024000
Encompass
NEB 5-alpha Competent E. coli (High Efficiency) is a derivative of the popular DH5. It is T1 phage resistant and endA deficient for high-quality plasmid preparations.
  • High efficiency strain ideal for cloning large plasmids and BACs
  • Also available in chemically-competent format (single-use and 200 µl vials)
  • No dry ice surcharge on competent cell shipments
  • Outgrowth medium included
  • Free of animal products
Catalog No. 50-202-4000 Supplier New England Biolabs, Inc. Supplier No. C2987R
NEB 5-alpha Competent E. coli (High Efficiency) is a derivative of the popular DH5. It is T1 phage resistant and endA deficient for high-quality plasmid preparations.
  • High efficiency strain ideal for cloning large plasmids and BACs
  • Also available in chemically-competent format (single-use and 200 µl vials)
  • No dry ice surcharge on competent cell shipments
  • Outgrowth medium included
  • Free of animal products