Learn More
New England Biolabs, Inc. NEB 5-alpha Competent E. coli (Subcloning Efficiency) – 6 x 0.4 ml/tube
SDP
Catalog No. 50994912
Encompass
Chemically competent E. coli cells suitable for subcloning efficiency transformations, such as plasmid transformation or routine subcloning (e.g,. inserting a 1 kb fragment into a 2 kb vector). The efficiency of these cells is not sufficiently high for most other cloning applications. For that, we recommend NEB 5-alpha Competent E. coli (High Efficiency) which has 1,000-fold higher transformation efficiency.
Catalog No. 50-994-912 Supplier New England Biolabs, Inc. Supplier No. C2988J
Chemically competent E. coli cells suitable for subcloning efficiency transformations, such as plasmid transformation or routine subcloning (e.g,. inserting a 1 kb fragment into a 2 kb vector). The efficiency of these cells is not sufficiently high for most other cloning applications. For that, we recommend NEB 5-alpha Competent E. coli (High Efficiency) which has 1,000-fold higher transformation efficiency.